CO1H324A619D CombiPhos Catalysts Inc

2-Chloro-3-cyclopropyl-5-methoxyphenylboronic acid

  • Cas Number : 2225176-26-9
  • Molecular Weight : 226.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart