CO1H324A61A1 CombiPhos Catalysts Inc

2-Fluoro-4-bromo-5-cyclopropylphenylboronic acid

  • Cas Number : 2225179-67-7
  • Molecular Weight : 258.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart