CO1H324A61A8 CombiPhos Catalysts Inc

4-Chloro-3-methoxy-6-bromophenylboronic acid

  • Cas Number : 2225176-63-4
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart