CO1H324A61B4 CombiPhos Catalysts Inc

3-Cyclopropyl-5-(iso-propyl)phenylboronic acid

  • Cas Number : 2225176-74-7
  • Molecular Weight : 204.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart