CO1H324A61B5 CombiPhos Catalysts Inc

3,5-Dicyclopropylphenylboronic acid

  • Cas Number : 2225172-67-6
  • Molecular Weight : 202.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart