CO1H324A61B9 CombiPhos Catalysts Inc

3-Cyclopropyl-5-(sec-butyl)phenylboronic acid

  • Cas Number : 2225152-93-0
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart