CO1H324A61BF CombiPhos Catalysts Inc

3-Trifluoromethyl-5-(thiazol-4-yl)phenylboronic acid

  • Cas Number : 2225181-02-0
  • Molecular Weight : 273.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart