CO1H324A61CA CombiPhos Catalysts Inc

3-Methyl-5-(piperidin-4-yl)phenylboronic acid

  • Cas Number : 2225178-66-3
  • Molecular Weight : 219.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart