CO1H324A61D0 CombiPhos Catalysts Inc

3-Bromo-4-(piperidin-4-yl)phenylboronic acid

  • Cas Number : 2225173-35-1
  • Molecular Weight : 283.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart