CO1H324A61D5 CombiPhos Catalysts Inc

(2-[1-[(TERT-BUTOXY)CARBONYL]PIPERIDIN-3-YL]PHENYL)BORONIC ACID

  • Cas Number : 2225152-38-3
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart