CO1H324A61D9 CombiPhos Catalysts Inc

(4-(piperidin-3-yl)-3-(trifluoromethyl)phenyl)boronic acid

  • Cas Number : 2256708-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart