CO1H324A61DB CombiPhos Catalysts Inc

2-Fluoro-5-(piperidin-3-yl)phenylboronic acid

  • Cas Number : 2225153-44-4
  • Molecular Weight : 223.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart