CO1H324A61DE CombiPhos Catalysts Inc

2-Methoxy-5-(piperidin-3-yl)phenylboronic acid

  • Cas Number : 2225152-06-5
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart