CO1H324A61E3 CombiPhos Catalysts Inc

2-(Piperidin-3-yl)-4-methylphenylboronic acid

  • Cas Number : 2225152-95-2
  • Molecular Weight : 219.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart