CO1H324A61E5 CombiPhos Catalysts Inc

3-(Piperidin-3-yl)-5-trifluoromethylphenylboronic acid

  • Cas Number : 2225152-66-7
  • Molecular Weight : 273.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart