CO1H324A61EF CombiPhos Catalysts Inc

4-(Piperidin-4-yl)-3-trifluoromethylphenylboronic acid

  • Cas Number : 2225169-45-7
  • Molecular Weight : 273.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart