CO1H324A61F6 CombiPhos Catalysts Inc

3-(Piperidin-4-yl)-4-methylphenylboronic acid

  • Cas Number : 2225173-10-2
  • Molecular Weight : 219.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart