CO1H324A61F7 CombiPhos Catalysts Inc

5-(Piperidin-4-yl)-2-aminophenylboronic acid

  • Cas Number : 2225181-04-2
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart