CO1H324A61FB CombiPhos Catalysts Inc

5-(Piperidin-4-yl)-2-cyanophenylboronic acid

  • Cas Number : 2225169-52-6
  • Molecular Weight : 230.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart