CO1H324A6201 CombiPhos Catalysts Inc

5-Ethylfuran-2-boronic acid

  • Cas Number : 162607-27-4
  • Molecular Weight : 139.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart