CO1H324A6202 CombiPhos Catalysts Inc

4-Ethylthiophene-2-boronic acid

  • Cas Number : 1142946-75-5
  • Molecular Weight : 156.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart