CO1H324A620A CombiPhos Catalysts Inc

5-(2-trifluoromethylphenyl)furan-2-boronic acid

  • Cas Number : 2096331-53-0
  • Molecular Weight : 255.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart