CO1H324A620F CombiPhos Catalysts Inc

3-(Piperidin-4-yl)-5-(iso-butyl)phenylboronic acid

  • Cas Number : 2225174-85-4
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart