CO1H324A6210 CombiPhos Catalysts Inc

3-(Cyclopropyl)-5-(iso-butyl)phenylboronic acid

  • Cas Number : 2225169-44-6
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart