CO1H324A6211 CombiPhos Catalysts Inc

4-(Cyclohexyl)thiophene-2-boronic acid

  • Cas Number : 2225181-06-4
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart