CO1H324A6214 CombiPhos Catalysts Inc

3-(tert-Butyl)-5-(pyridin-3-yl)phenylboronic acid

  • Cas Number : 2225173-54-4
  • Molecular Weight : 255.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart