CO1H324A6216 CombiPhos Catalysts Inc

3-(tert-Butyl)-5-(pyrimidin-5-yl)phenylboronic acid

  • Cas Number : 2225170-29-4
  • Molecular Weight : 256.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart