CO1H324A6219 CombiPhos Catalysts Inc

4-(3-Tolyl)thiophene-2-boronic acid

  • Cas Number : 2122282-61-3
  • Molecular Weight : 218.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart