CO1H324A621F CombiPhos Catalysts Inc

3-Cyclopropyl-5-(3-methylpiperidin-1-yl)phenylboronic acid

  • Cas Number : 2225152-82-7
  • Molecular Weight : 259.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart