CO1H324A6230 CombiPhos Catalysts Inc

2-Fluoro-4-(sec-butyl)phenylboronic acid

  • Cas Number : 2225152-55-4
  • Molecular Weight : 196.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart