CO1H324A6235 CombiPhos Catalysts Inc

2-Fluoro-4-(cyclohexyl)phenylboronic acid

  • Cas Number : 2225179-99-5
  • Molecular Weight : 222.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart