CO1H324A624C CombiPhos Catalysts Inc

3-Cyclopropyl-5-(4-methylimidazol-1-yl)phenylboronic acid

  • Cas Number : 2225170-82-9
  • Molecular Weight : 242.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart