CO1H324A624D CombiPhos Catalysts Inc

4-(4-Methoxyphenyl)thiophene-2-boronic acid

  • Cas Number : 2225176-86-1
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart