CO1H324A624E CombiPhos Catalysts Inc

3,5-Bis(pyrrolidino)phenylboronic acid

  • Cas Number : 2225170-56-7
  • Molecular Weight : 260.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart