CO1H324A6251 CombiPhos Catalysts Inc

3,5-Bis(1H-pyrrol-1-yl)phenylboronic acid

  • Cas Number : 2225172-18-7
  • Molecular Weight : 252.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart