CO1H324A6255 CombiPhos Catalysts Inc

4-(Cyclopentyl)furan-2-boronic acid

  • Cas Number : 2225155-72-4
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart