CO1H324A6256 CombiPhos Catalysts Inc

4-(Pyrrolidin-3-yl)phenylboronic acid

  • Cas Number : 2225152-56-5
  • Molecular Weight : 191.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart