CO1H324A625B CombiPhos Catalysts Inc

(4'-FLUORO-[1,1'-BIPHENYL]-2-YL)BORONIC ACID

  • Cas Number : 179526-94-4
  • Molecular Weight : 216.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart