CO1H324A6262 CombiPhos Catalysts Inc

4-(4-Tolyl)furan-2-boronic acid

  • Cas Number : 2096331-45-0
  • Molecular Weight : 202.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart