CO1H324A626C CombiPhos Catalysts Inc

4-(3-Fluorophenyl)furan-2-boronic acid

  • Cas Number : 2096334-71-1
  • Molecular Weight : 205.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart