CO1H324A626D CombiPhos Catalysts Inc

5-Bromo-2-(1H-imidazol-1-yl)phenylboronic acid

  • Cas Number : 2225173-63-5
  • Molecular Weight : 266.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart