CO1H324A6271 CombiPhos Catalysts Inc

3-Bromo-4-(1H-pyrazol-1-yl)phenylboronic acid

  • Cas Number : 2225177-78-4
  • Molecular Weight : 266.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart