CO1H324A6276 CombiPhos Catalysts Inc

2-(Morpholin-2-yl)phenylboronic acid

  • Cas Number : 2225153-33-1
  • Molecular Weight : 207.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart