CO1H324A6277 CombiPhos Catalysts Inc

3-(Morpholin-2-yl)phenylboronic acid

  • Cas Number : 2225152-23-6
  • Molecular Weight : 207.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart