CO1H324A6283 CombiPhos Catalysts Inc

(6-propylpyridin-3-yl)boronic acid

  • Cas Number : 1201788-51-3
  • Molecular Weight : 165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart