CO1H324A6289 CombiPhos Catalysts Inc

(6-cyclohexylpyridin-3-yl)boronic acid

  • Cas Number : 2225173-72-6
  • Molecular Weight : 205.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart