CO1H324A628C CombiPhos Catalysts Inc

(6-(thiophen-2-yl)pyridin-3-yl)boronic acid

  • Cas Number : 2225169-76-4
  • Molecular Weight : 205.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart