CO1H324A628E CombiPhos Catalysts Inc

2-(Cyclohexeyl)pyridine-5-boronic acid

  • Cas Number : 1264514-93-3
  • Molecular Weight : 203.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart