CO1H324A6290 CombiPhos Catalysts Inc

2-(4-Ethylphenyl)pyridine-4-boronic acid

  • Cas Number : 2225172-29-0
  • Molecular Weight : 227.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart