CO1H324A6293 CombiPhos Catalysts Inc

2-(2-HYDROXYPROPAN-2-YL)PYRIDINE-4-BORONIC ACID

  • Cas Number : 2225156-03-4
  • Molecular Weight : 181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart